Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128949.3 | NP_565776.1 | 11 - 12 | 1649 | 105 | |
GenBank mRNA | AF360290.1 | - | 11 - 12 | 1571 | 105 | |
AK175540.1 | - | 11 - 12 | 1551 | 105 | ||
AK229595.1 | - | 9 - 10 | 3166 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |