Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336477.1 | NP_001325145.1 | 5 - 6 | 1025 | 130 | |
NM_128949.3 | NP_565776.1 | 5 - 6 | 929 | 130 | ||
GenBank mRNA | AF360290.1 | - | 5 - 6 | 1001 | 130 | |
AK175540.1 | - | 5 - 6 | 927 | 130 | ||
AK229595.1 | - | 5 - 6 | 972 | 130 | ||
AY051046.1 | - | 5 - 6 | 868 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons |