Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036404.1 | NP_001031481.1 | 3 - 4 | 417 | 99 | |
NM_128999.3 | NP_180994.1 | 3 - 4 | 414 | 99 | ||
GenBank mRNA | AK119138.1 | - | 3 - 4 | 379 | 99 | |
AY085812.1 | - | 3 - 4 | 414 | 99 | ||
BT005276.1 | - | 3 - 4 | 320 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |