Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036404.1 | NP_001031481.1 | 5 - 6 | 732 | 60 | |
NM_128999.3 | NP_180994.1 | 5 - 6 | 729 | 60 | ||
GenBank mRNA | AK119138.1 | - | 5 - 6 | 694 | 60 | |
AY085812.1 | - | 5 - 6 | 729 | 60 | ||
BT005276.1 | - | 5 - 6 | 635 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |