Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036094.1 | NP_001031171.1 | 12 - 13 | 1474 | 99 | |
NM_104037.4 | NP_175570.1 | 12 - 13 | 1478 | 99 | ||
GenBank mRNA | AY081353.1 | - | 12 - 13 | 1465 | 99 | |
AY128845.1 | - | 12 - 13 | 1292 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |