Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036094.1 | NP_001031171.1 | 13 - 14 | 1648 | 159 | |
NM_104037.4 | NP_175570.1 | 13 - 14 | 1652 | 159 | ||
GenBank mRNA | AY081353.1 | - | 13 - 14 | 1639 | 159 | |
AY128845.1 | - | 13 - 14 | 1466 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |