Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104038.4 | NP_564593.1 | 2 - 3 | 773 | 88 | |
NM_202273.3 | NP_974002.1 | 2 - 3 | 535 | 88 | ||
GenBank mRNA | AB119061.1 | - | 2 - 3 | 447 | 88 | |
AY150395.1 | - | 1 - 2 | 306 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |