Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198270.1 | NP_001185199.1 | 5 - 6 | 629 | 127 | |
NM_001333494.1 | NP_001322512.1 | 3 - 4 | 2525 | 127 | ||
NM_001333495.1 | NP_001322511.1 | 5 - 6 | 647 | 127 | ||
NM_104047.4 | NP_564595.1 | 4 - 5 | 2254 | 127 | ||
NM_202274.2 | NP_974003.1 | 3 - 4 | 551 | 127 | ||
GenBank mRNA | AK230310.1 | - | 4 - 5 | 2034 | 127 | |
AY120756.1 | - | 3 - 4 | 549 | 127 | ||
BT000109.1 | - | 3 - 4 | 307 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |