Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATHB-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084233.2 | NP_001077702.1 | 8 - 9 | 1810 | 76 | |
NM_104096.3 | NP_175627.1 | 8 - 9 | 1810 | 76 | ||
NM_179464.1 | NP_849795.1 | 8 - 9 | 1724 | 76 | ||
GenBank mRNA | AJ439449.1 | - | 8 - 9 | 1007 | 76 | |
AY060556.1 | - | 8 - 9 | 1724 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |