Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084854.2 | NP_001078323.1 | 3 - 4 | 879 | 74 | |
NM_116008.4 | NP_191702.1 | 2 - 3 | 1036 | 74 | ||
GenBank mRNA | AK229222.1 | - | 2 - 3 | 689 | 74 | |
AY058113.1 | - | 2 - 3 | 689 | 74 | ||
AY097398.1 | - | 2 - 3 | 622 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |