Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084855.2 | NP_001078324.1 | 2 - 3 | 296 | 97 | |
NM_001340099.1 | NP_001327883.1 | 3 - 4 | 472 | 97 | ||
NM_116009.4 | NP_191703.1 | 3 - 4 | 540 | 97 | ||
GenBank mRNA | AB024282.1 | - | 3 - 4 | 388 | 97 | |
AJ010505.1 | - | 3 - 4 | 417 | 97 | ||
AK226606.1 | - | 3 - 4 | 410 | 97 | ||
AY087208.1 | - | 3 - 4 | 472 | 97 | ||
AY093094.1 | - | 3 - 4 | 474 | 97 | ||
AY128782.1 | - | 3 - 4 | 334 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |