Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035837.1 | NP_001030914.1 | 8 - 9 | 1582 | 131 | |
NM_116036.4 | NP_567116.1 | 9 - 10 | 1500 | 131 | ||
NM_202746.2 | NP_974475.1 | 8 - 9 | 1590 | 131 | ||
GenBank mRNA | AY039613.1 | - | 9 - 10 | 1486 | 131 | |
BT000508.1 | - | 9 - 10 | 1307 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |