Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084857.2 | NP_001078326.1 | 16 - 17 | 2834 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |