Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084857.2 | NP_001078326.1 | 9 - 10 | 2004 | 141 | |
NM_116039.4 | NP_191733.3 | 9 - 10 | 1960 | 141 | ||
GenBank mRNA | AB493660.1 | - | 9 - 10 | 1536 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |