Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340129.1 | NP_001327870.1 | 5 - 6 | 737 | 116 | |
NM_116048.3 | NP_567119.1 | 6 - 7 | 845 | 116 | ||
GenBank mRNA | AF334717.1 | - | 6 - 7 | 563 | 116 | |
AY059746.1 | - | 6 - 7 | 783 | 116 | ||
AY070463.1 | - | 6 - 7 | 848 | 116 | ||
AY091392.1 | - | 6 - 7 | 563 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |