Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340138.1 | NP_001327812.1 | 1 - 2 | 422 | 82 | |
NM_116075.3 | NP_191769.1 | 1 - 2 | 513 | 82 | ||
GenBank mRNA | AB493662.1 | - | 1 - 2 | 364 | 82 | |
AY669801.1 | - | 1 - 2 | 364 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |