Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZIP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116107.2 | NP_191801.1 | 1 | 960 | 100 | |
GenBank mRNA | AF400620.1 | - | 1 | 394 | 100 | |
AY050923.1 | - | 1 | 920 | 100 | ||
AY091421.1 | - | 1 | 394 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |