Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116125.3 | NP_191819.1 | 5 - 6 | 886 | 78 | |
GenBank mRNA | AY087640.1 | - | 5 - 6 | 747 | 78 | |
AY094429.1 | - | 5 - 6 | 716 | 78 | ||
AY125534.1 | - | 5 - 6 | 583 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |