Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116125.3 | NP_191819.1 | 7 - 8 | 1032 | 66 | |
GenBank mRNA | AY087640.1 | - | 7 - 8 | 893 | 66 | |
AY094429.1 | - | 7 - 8 | 862 | 66 | ||
AY125534.1 | - | 7 - 8 | 729 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |