Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116125.3 | NP_191819.1 | 8 - 9 | 1119 | 103 | |
GenBank mRNA | AY087640.1 | - | 8 - 9 | 980 | 103 | |
AY094429.1 | - | 8 - 9 | 949 | 103 | ||
AY125534.1 | - | 8 - 9 | 816 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |