Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116135.3 | NP_191829.1 | 6 - 7 | 4412 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |