Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOEP16-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340181.1 | NP_001319822.1 | 1 - 2 | 263 | 58 | |
NM_116153.5 | NP_191847.1 | 2 - 3 | 192 | 58 | ||
GenBank mRNA | AK117583.1 | - | 2 - 3 | 132 | 58 | |
BT005150.1 | - | 2 - 3 | 128 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |