Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOEP16-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340181.1 | NP_001319822.1 | 2 - 3 | 322 | 80 | |
NM_116153.5 | NP_191847.1 | 3 - 4 | 251 | 80 | ||
GenBank mRNA | AK117583.1 | - | 3 - 4 | 191 | 80 | |
BT005150.1 | - | 3 - 4 | 187 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |