Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATOEP16-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340181.1 | NP_001319822.1 | 3 - 4 | 545 | 100 | |
NM_116153.5 | NP_191847.1 | 4 - 5 | 474 | 100 | ||
GenBank mRNA | AK117583.1 | - | 4 - 5 | 414 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |