Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116156.5 | NP_191850.2 | 8 - 9 | 726 | 74 | |
GenBank mRNA | AB109893.1 | - | 8 - 9 | 632 | 74 | |
AY090253.1 | - | 8 - 9 | 673 | 74 | ||
AY125535.1 | - | 8 - 9 | 632 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |