Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116184.4 | NP_567141.1 | 3 - 4 | 454 | 129 | |
GenBank mRNA | AF372883.1 | - | 3 - 4 | 342 | 129 | |
AY056778.1 | - | 3 - 4 | 422 | 129 | ||
AY092982.1 | - | 3 - 4 | 360 | 129 | ||
BT002678.1 | - | 3 - 4 | 293 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |