Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116191.3 | NP_191885.1 | 2 - 3 | 1096 | 64 | |
NM_202754.2 | NP_974483.1 | 2 - 3 | 1029 | 64 | ||
GenBank mRNA | AB006810.1 | - | 2 - 3 | 697 | 64 | |
AY080717.1 | - | 2 - 3 | 646 | 64 | ||
AY117337.1 | - | 2 - 3 | 438 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |