Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125427.1 | NP_001118899.1 | 13 - 14 | 882 | 85 | |
NM_001340197.1 | NP_001326587.1 | 14 - 15 | 921 | 85 | ||
NM_001340198.1 | NP_001326588.1 | 13 - 14 | 921 | 85 | ||
NM_116193.5 | NP_191887.2 | 14 - 15 | 962 | 85 | ||
GenBank mRNA | AY080682.1 | - | 14 - 15 | 956 | 85 | |
AY117234.1 | - | 12 - 13 | 745 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |