Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116217.3 | NP_191911.1 | 11 - 12 | 1361 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |