Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340229.1 | NP_001328569.1 | 14 - 15 | 4383 | 84 | |
NM_001340230.1 | NP_001328568.1 | 14 - 15 | 4382 | 84 | ||
NM_116223.4 | NP_191917.3 | 14 - 15 | 4402 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |