Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340229.1 | NP_001328569.1 | 8 - 9 | 3751 | 66 | |
NM_001340230.1 | NP_001328568.1 | 8 - 9 | 3750 | 66 | ||
NM_116223.4 | NP_191917.3 | 8 - 9 | 3770 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |