Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340236.1 | NP_001328541.1 | 6 - 7 | 1093 | 93 | |
NM_116240.5 | NP_567155.1 | 6 - 7 | 1244 | 93 | ||
GenBank mRNA | AF190794.1 | - | 6 - 7 | 1064 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |