Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340239.1 | NP_001329075.1 | 3 - 4 | 792 | 71 | |
NM_116255.5 | NP_567170.2 | 3 - 4 | 756 | 71 | ||
GenBank mRNA | AK227113.1 | - | 3 - 4 | 722 | 71 | |
AY064064.1 | - | 3 - 4 | 720 | 71 | ||
AY133754.1 | - | 3 - 4 | 622 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |