Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116261.4 | NP_567175.2 | 1 - 2 | 299 | 95 | |
GenBank mRNA | AK118390.1 | - | 1 - 2 | 174 | 95 | |
BT009663.1 | - | 1 - 2 | 53 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |