Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125431.2 | NP_001118903.1 | 2 - 3 | 580 | 117 | |
NM_001340252.1 | NP_001328184.1 | 1 - 2 | 488 | 117 | ||
NM_001340253.1 | NP_001328183.1 | 1 - 2 | 508 | 117 | ||
NM_116271.3 | NP_191956.2 | 2 - 3 | 580 | 117 | ||
GenBank mRNA | BT030385.1 | - | 1 - 2 | 317 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |