Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160722.2 | NP_001154194.1 | 6 - 7 | 1195 | 76 | |
NM_001340255.1 | NP_001329692.1 | 6 - 7 | 1231 | 76 | ||
NM_116272.4 | NP_191957.2 | 6 - 7 | 1340 | 76 | ||
GenBank mRNA | AB493664.1 | - | 6 - 7 | 1098 | 76 | |
AF251697.1 | - | 6 - 7 | 1098 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |