Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116297.4 | NP_191981.1 | 2 - 3 | 785 | 74 | |
GenBank mRNA | AK226199.1 | - | 2 - 3 | 604 | 74 | |
AY062099.1 | - | 2 - 3 | 597 | 74 | ||
AY124881.1 | - | 2 - 3 | 308 | 74 | ||
Y07822.1 | - | 506 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |