Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116297.4 | NP_191981.1 | 5 - 6 | 1213 | 107 | |
GenBank mRNA | AK226199.1 | - | 5 - 6 | 1032 | 107 | |
AY062099.1 | - | 5 - 6 | 1025 | 107 | ||
AY124881.1 | - | 5 - 6 | 736 | 107 | ||
Y07822.1 | - | 934 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |