Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125435.1 | NP_001118907.1 | 4 - 5 | 1593 | 94 | |
NM_116298.4 | NP_567183.2 | 4 - 5 | 1697 | 94 | ||
GenBank mRNA | AK226968.1 | - | 4 - 5 | 1538 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |