Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116342.3 | NP_192021.1 | 3 - 4 | 295 | 112 | |
GenBank mRNA | AY062667.1 | - | 3 - 4 | 253 | 112 | |
AY093316.1 | - | 2 - 3 | 112 | 112 | ||
X63895.1 | - | 197 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |