Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340304.1 | NP_001329151.1 | 1 - 2 | 385 | 119 | |
NM_001340305.1 | NP_001329152.1 | 2 - 3 | 269 | 119 | ||
NM_116349.1 | NP_192028.1 | 1 - 2 | 113 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |