Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340304.1 | NP_001329151.1 | 8 - 9 | 1195 | 66 | |
NM_001340305.1 | NP_001329152.1 | 9 - 10 | 1079 | 66 | ||
NM_116349.1 | NP_192028.1 | 8 - 9 | 923 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |