Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116367.3 | NP_192046.1 | 4 - 5 | 1081 | 88 | |
GenBank mRNA | AF360231.1 | - | 4 - 5 | 828 | 88 | |
AY040031.1 | - | 4 - 5 | 779 | 88 | ||
AY088537.1 | - | 4 - 5 | 903 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |