Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340322.1 | NP_001328722.1 | 4 - 5 | 326 | 80 | |
NM_001340323.1 | NP_001328723.1 | 4 - 5 | 326 | 80 | ||
NM_001340324.1 | NP_001328724.1 | 4 - 5 | 315 | 80 | ||
NM_001340325.1 | NP_001328720.1 | 3 - 4 | 403 | 80 | ||
NM_001340326.1 | NP_001328725.1 | 4 - 5 | 307 | 80 | ||
NM_001340327.1 | NP_001328721.1 | 3 - 4 | 388 | 80 | ||
NM_001340328.1 | NP_001328726.1 | 2 - 3 | 238 | 80 | ||
NM_116381.4 | NP_192060.2 | 4 - 5 | 326 | 80 | ||
GenBank mRNA | AY758071.1 | - | 4 - 5 | 307 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |