Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116386.4 | NP_567213.1 | 4 - 5 | 619 | 140 | |
GenBank mRNA | AY050341.1 | - | 4 - 5 | 551 | 140 | |
AY086463.1 | - | 4 - 5 | 572 | 140 | ||
BT020609.1 | - | 4 - 5 | 507 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |