Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116396.4 | NP_192075.1 | 2 - 3 | 1064 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |