Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116402.4 | NP_192081.1 | 1 - 2 | 365 | 103 | |
GenBank mRNA | AF480165.1 | - | 1 - 2 | 220 | 103 | |
AK229522.1 | - | 1 - 2 | 248 | 103 | ||
BT008704.1 | - | 1 - 2 | 248 | 103 | ||
BT029024.1 | - | 1 - 2 | 220 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |