Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340350.1 | NP_001328930.1 | 2 - 3 | 2130 | 81 | |
NM_116428.5 | NP_192106.3 | 2 - 3 | 1749 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |