Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340367.1 | NP_001328074.1 | 4 - 5 | 630 | 68 | |
NM_116447.3 | NP_192124.1 | 4 - 5 | 630 | 68 | ||
GenBank mRNA | AF385693.1 | - | 4 - 5 | 629 | 68 | |
AY081725.1 | - | 4 - 5 | 542 | 68 | ||
DQ087970.1 | - | 4 - 5 | 542 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |