Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340369.1 | NP_001329795.1 | 8 - 9 | 1186 | 73 | |
NM_116459.4 | NP_567226.1 | 8 - 9 | 1203 | 73 | ||
NM_178956.4 | NP_849287.2 | 8 - 9 | 1194 | 73 | ||
NM_202772.2 | NP_974501.1 | 8 - 9 | 1194 | 73 | ||
GenBank mRNA | AF225702.1 | - | 8 - 9 | 1047 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |